missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Antibody (6A5F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 217.00 - € 557.00
Specifications
| Antigen | BNIP3 |
|---|---|
| Clone | 6A5F7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214844
|
Novus Biologicals
NBP2-61715 |
0.1 mL |
€ 557.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678967
|
Novus Biologicals
NBP2-61715-0.025ml |
0.025 mL |
€ 217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BNIP3 Monoclonal specifically detects BNIP3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| BNIP3 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors | |
| 664 | |
| Purified recombinant fragment of human BNIP3 (AA: 50-155) expressed in E. Coli. | |
| Primary | |
| 21.5 kDa |
| 6A5F7 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Unconjugated | |
| Mouse | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
| BNIP3 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title