missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIPL Monoclonal Antibody (OTI8F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809281S
This item is not returnable.
View return policy
Description
BNIPL Monoclonal antibody specifically detects BNIPL in Human samples. It is validated for Western BlotSpecifications
| BNIPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z465 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 149428 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8F6 | |
| Unconjugated | |
| BNIPL | |
| BCL2 interacting protein like; Bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein; BCL2/adenovirus E1B 19kD interacting protein like; BNIP-2 similar; BNIPL; BNIPL1; BNIPL-1; BNIPL2; BNIPL-2; BNIPS; BNIP-S; BNIP-Salpha; BNIP-Sbeta; PP753 | |
| Human recombinant protein fragment corresponding to amino acids 1-203 of human BNIPL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction