Learn More
BNIPL Monoclonal Antibody (OTI9C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809280S
This item is not returnable.
View return policy
Description
BNIPL Monoclonal antibody specifically detects BNIPL in Human samples. It is validated for Western BlotSpecifications
| BNIPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q08431 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4240 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9C1 | |
| Unconjugated | |
| MFGE8 | |
| BCL2 interacting protein like; Bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein; BCL2/adenovirus E1B 19kD interacting protein like; BNIP-2 similar; BNIPL; BNIPL1; BNIPL-1; BNIPL2; BNIPL-2; BNIPS; BNIP-S; BNIP-Salpha; BNIP-Sbeta; PP753 | |
| Full length human recombinant protein of human MFGE8 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |