Learn More
Emma Safety Footwear Bob XXD Safety Shoes with Sturdy Steel Toe
Product Code. 13244384
Change view
Click to view available options
Size:
43
45
46
Unit Size:
1 pair
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
13244384 43 1 pair
11373363 43 1 pair
11362770 45 Each
11303110 46 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13244384 Supplier Emma Safety Footwear Supplier No. 615036/43
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Vibram™-designed sole highly resistant to extreme temperatures and corrosive chemicals.
  • Full grain leather uppers
  • Hydro-Tec™ sanitized silver lining
  • Woven zero penetration insole
  • GripForce™ Easy Twist/XXD Vibram outsole
Specifications
Color Black
Liner Material Hydro-Tec Sanitized Silver lining
Material Leather
Size 43
Toe Material Steel
Product Type Safety Shoe