missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOB1 Antibody (BOB1/2423), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08666AF488
This item is not returnable.
View return policy
Description
BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| BOB1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
| Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) (Uniprot: Q16633) | |
| 100 μg | |
| Immunology | |
| 5450 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| BOB1/2423 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction