missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOB1 Antibody (OTI1E11), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71223AF350
This item is not returnable.
View return policy
Description
BOB1 Monoclonal specifically detects BOB1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| BOB1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| POU2AF1 | |
| Full length recombinant protein of human BOB1 (NP_006226) produced in HEK293T cells. | |
| Immunology | |
| 5450 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1E11 | |
| Western Blot | |
| B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction