missing translation for 'onlineSavingsMsg'
Learn More

BOB1 Antibody (SP92), Novus Biologicals™

Product Code. p-200132274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
1 mL
Unit Size:
0.1mL
0.50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532815 1 mL -
30532814 0.5 mL 0.50mL
30532813 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30532815 Supplier Novus Biologicals Supplier No. NBP3451091ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOB1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP92
Conjugate Unconjugated
Formulation PBS (pH 7.4), Tween-20, 0.2% BSA
Gene Alias B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1
Host Species Rabbit
Immunogen Synthetic peptide within Human BOB1 aa 200 to the C-terminus (C terminal). The exact sequence is proprietary
Purification Method Protein A purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5450
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.