Learn More
Boekel Scientific™ InSlide Out™ Hybridization Apparatus Polycarbonate Slide Holders
Product Code. 10069832
Click to view available options
Material:
Polycarbonate
Unit Size:
Each
Product Code. 10069832
Brand: Boekel Scientific™ C2403765
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Holds up to eighteen 1 x 3 in. slides only
  • Holds up to eighteen 1 × 3 in. slides only
  • Slides are placed in slots for ease in carrying
Specifications
For Use With (Equipment) InSlide out hybridization apparatus
Holds Eighteen 1 x 3 in. slide
Material Polycarbonate
Type Slide Holder
For Use With (Application) InSlide out hybridization apparatus