missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOK Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41045IR
This item is not returnable.
View return policy
Description
BOK Polyclonal antibody specifically detects BOK in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BOK | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BOK. The immunogen is located within the first 50 amino acids of BOK. Amino Acid Squence: DAFDRSPTDKELVAQA | |
| 0.1 mL | |
| Apoptosis | |
| 666 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction