Learn More
Bohlender™ BOLA™ Measuring Cylinders
Product Code. p-7237730
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Graduations:
1 mL
2 mL
2.5 mL
5 mL
Diameter (Metric) Inside:
24 mm
30 mm
40 mm
53 mm
Height (Metric):
172 mm
202 mm
268 mm
308 mm
Unit Size:
Each
Product Code. 17186924
Brand: Bohlender™ A16412
Translucent measuring cylinders with hexagonal base to prevent rolling.
  • Translucent and non-porous
  • Design based on ISO 4788
  • With graduation (uncalibrated), spout and reinforcing ring
  • Universal chemical resistance
  • Product is only exposed to PTFE
  • Hexagonal base prevents the cylinder from rolling
  • Permanent, recessed graduation
Specifications
Capacity (Metric) 50 mL
Material PTFE
Graduations 1 mL
Capacity (English) 1.7 oz.
Shape Cylinder
Diameter (Metric) Inside 24 mm
Diameter (English) Inside 0.94 in.
Height (English) 6.8 in.
Height (Metric) 172 mm