Learn More
Bohlender™ Bola PTFE/PEEK Propeller with 4 Blades
Click to view available options
Diameter (Metric) Shaft:
8 mm
10 mm
Dimensions (L x W x H):
100 L x 25 W x 5 Hmm
50 L x 22 W x 4 Hmm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Shaft | 8 mm |
| For Use With (Application) | The product is sucked bottom-up, good axial flow with low shear force. |
| Includes | 4 Blades |
| Quantity | 1 Each |
| Temperature (Metric) | -200°C to +250°C |
| Type | Propeller |
| Dimensions (L x W x H) | 50 L x 22 W x 4 Hmm |