Learn More
Bohlender™ BOLA™ PTFE Stirrer Bearings
Click to view available options
Length (Metric):
65mm
70mm
72mm
74mm
86mm
Diameter (Metric) Shaft:
6 mm
8 mm
10 mm
16 mm
Unit Size:
Each
Description
- Ground joint cone made of PTFE with sealing rings on the outside to prevent sticking of the connection and to reduce danger of breaking glass
- Suitable for vacuum
- Universal chemical resistance
- Temperature range: -15 to +200°C
Specifications
Specifications
| Diameter (Metric) Shaft | 8 mm |
| For Use With (Application) | Suitable for vacuum, perfect bearing for Stainless Steel, glass and BOLA Stirrer Shafts |
| Length (Metric) | 74mm |
| Quantity | 1 Ea. |
| Standard Taper Joint | 29/32 |
| Temperature (Metric) | -200°C to +250°C |
| Type | Stirrer Bearing |
| Material | PTFE |