missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99968MFV500
This item is not returnable.
View return policy
Description
BOLA1 Polyclonal antibody specifically detects BOLA1 in Human samples. It is validated for ELISA
Specifications
| BOLA1 | |
| mFluor Violet 500 SE | |
| Antigen and protein A Affinity-purified | |
| Store at 4C in the dark. |
| ELISA | |
| Produced in rabbits immunized with purified, recombinant Human BOLA1 (Uniprot#: Q9Y3E2; Met1-Pro137). | |
| 0.1 mL | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction