missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA2 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18507JF669
This item is not returnable.
View return policy
Description
BOLA2 Polyclonal antibody specifically detects BOLA2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| BOLA2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 654483 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 669 | |
| BolA Homolog 2B (E. Coli), BOLA2, BOLA2A, BOLA2B, BolA-Like 2B, BolA-Like 2B (E. Coli), BolA-Like Protein 2, BolA-Like Protein 2 Member B, BolA-Like Protein 2B | |
| Recognizes region between residues Between between 36 and 86.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction