missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA3 Antibody (PCRP-BOLA3-1A5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20915F
This item is not returnable.
View return policy
Description
BOLA3 Monoclonal antibody specifically detects BOLA3 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation
Specifications
| BOLA3 | |
| Monoclonal | |
| FITC | |
| bolA homolog 3 (E. coli), bolA-like 3, bolA-like protein 3 | |
| Recombinant fragment (around aa1-103) of human BOLA3 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 388962 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation | |
| PCRP-BOLA3-1A5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction