missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOLA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120414
This item is not returnable.
View return policy
Description
Positive test controls include: mouse stomach, mouse heart, mouse kidney, rat liver. The target is usually found in the following locations: Secreted. Immunogen sequence: MAAWSPAAAA PLLRGIRGLP LHHRMFATQT EGELRVTQIL KEKFPRATAI KVTDISGGCG AMYEIKIE.
This gene encodes a protein that plays an essential role in the production of iron-sulfur (Fe-S) clusters for the normal maturation of lipoate-containing 2-oxoacid dehydrogenases, and for the assembly of the mitochondrial respiratory chain complexes. Mutation in this gene has been associated with multiple mitochondrial dysfunctions syndrome-2. Two alternatively spliced transcript variants encoding different isoforms with distinct subcellular localization have been reported for this gene.
Specifications
| BOLA3 | |
| Polyclonal | |
| Unconjugated | |
| BOLA3 | |
| bolA family member 3; bolA homolog 3; BOLA3; bolA-like protein 3; MMDS2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 297388, 388962, 78653 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q53S33, Q8CEI1 | |
| BOLA3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-68 of human BOLA3 (NP_997717.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction