missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boll Antibody (2E8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00066037-M06
This item is not returnable.
View return policy
Description
Boll Monoclonal antibody specifically detects Boll in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Boll | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| bol, boule-like (Drosophila), BOULEbol (Drosophila boule homolog)-like, protein boule-like | |
| BOLL (NP_149019, 185 a.a. ∽ 283 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.ATTQYLPGQWQWSVPQPSASSAPFLYLQPSEVIYQPVEIAQDGGCVPPPLSLMETSVPEPYSDHGVQATYHQVYAPSAITMPAPVMQPEPIKTVWSIHY | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2E8 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NP_149019 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 66037 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction