missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bone marrow stromal cell antigen 1/CD157 Antibody (029), Alexa Fluor™ 700 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89249AF700
This item is not returnable.
View return policy
Description
Bone marrow stromal cell antigen 1/CD157 Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1/CD157 in Human samples. It is validated for ELISA
Specifications
| Bone marrow stromal cell antigen 1/CD157 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Bone marrow stromal cell antigen 1/CD157 (Met 1-Lys 292; NP_004325.2). | |
| 0.1 mL | |
| Immunology | |
| 683 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 029 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction