missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bone marrow stromal cell antigen 1/CD157 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-14363-25ul
This item is not returnable.
View return policy
Description
Bone marrow stromal cell antigen 1/CD157 Polyclonal specifically detects Bone marrow stromal cell antigen 1/CD157 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Bone marrow stromal cell antigen 1/CD157 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BST1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: EGTSAHLRDIFLGRCAEYRALLSPEQRNKNCTAIWEAFKVALDKDPCSVLPSDYDLFINLSRHSIPRDKSLFWE | |
| 25ul | |
| Immunology | |
| 683 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction