missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOP1 Antibody (T7-P1B7*A8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50124H
This item is not returnable.
View return policy
Description
BOP1 Monoclonal specifically detects BOP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BOP1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| BOP1 | |
| Peptide Sequence - KGHVLTRDLG | |
| 0.1 mL | |
| Epigenetics | |
| 23246 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| T7-P1B7*A8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| block of proliferation 1, KIAA0124Block of proliferation 1 protein, ribosome biogenesis protein BOP1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction