missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bora Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583521
This item is not returnable.
View return policy
Description
Immunogen sequence: RKFTVHSPDA SSGTNSNGIT NPCIRSPYID GCSPIKNWSP MRLQMYSGGT QYRTSVIQIP FTLETQGEDE EDKENIPSTD VSSPAMDAAG IHLRQF Highest antigen sequence indentity to the following orthologs: Mouse - 49%, Rat - 60%.
Bora (Protein aurora borealis) is a key activator of Aurora Related Protein Kinase A (ARK-1), which is a centrosome-associated serine/threonine kinase that regulates centrosome maturation, bipolar spindle assembly and chromosome segregation during mitosis. Bora is localized to the nucleus until mitosis is initiated, then translocates to the cytoplasm in a Cdc2 dependent manner. Activation of Cdc2 initiates the release of Bora into the cytoplasm where it can bind and activate ARK-1. PLK1 (polo-like kinase-1) interacts with Bora to control the accessibility of its activation loop for phosphorylation and activation by ARK-1. Bora and ARK-1 cooperatively activate PLK1 and control mitotic entry. Bora mutants result in multipolar spindles in mitosis identical to those observed when ARK-1 function is blocked. Thus, the ARK1-Bora-PLK1 regulatory circuit in mammalian cells elucidates a key mechanism in cell cycle regulation. At least three isoforms of Bora are known to exist.
Specifications
| Bora | |
| Polyclonal | |
| Unconjugated | |
| BORA | |
| 6720463M24Rik; AI317232; aurora borealis; BORA; bora, aurora kinase A activator; C13orf34; HsBora; Protein aurora borealis; RGD1309522; RP11-342J4.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PGQ7 | |
| BORA | |
| Recombinant protein corresponding to Human BORA. Recombinant protein control fragment (Product #RP-97972). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction