missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethyl sulfide complex, 2M in toluene, packaged under Argon in resealable ChemSeal™ bottles

Product Code. p-7058658
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11340248

Brand: Thermo Scientific Alfa Aesar 042963.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

Borane-dimethyl sulfide complex is used for hydroborations and reductions. Used with a dendrimeric supported L-pyrrolidinol in the asymmetric reduction of indanones and tetralones. CBS-catalyzed asymmetric reduction of ferrocenyl-1,3-diketones to 1,3-diols. Highly enantioselective reduction of ketones catalyzed by C3-symmetric tripodal hydroxyamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-87-0
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name boron;methylsulfanylmethane
SMILES B.CSC

Specifications

Color Colorless
Density 0.856 g/mL
Flash Point 7°C (45°F)
Packaging ChemSeal™ bottles
Linear Formula (CH3)2S·BH3
Quantity 100 mL
UN Number UN3399
Beilstein 3663489
Sensitivity Air and moisture sensitive
Solubility Information It reacts with water. Miscible with diethyl ether,methylene chloride,hexane,heptane,petroleum ether,tetrahydrofuran,diglyme,benzene,toluene and xylene.
Formula Weight 75.97
Concentration or Composition (by Analyte or Components) 2M in toluene
Physical Form Liquid
Chemical Name or Material Borane-dimethyl sulfide complex
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.