missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane dimethyl sulfide complex, 94%, AcroSeal™, Thermo Scientific Chemicals
139.50 EUR valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Use promo code "23251" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Melting Point | -40°C to 37°C |
Color | Amber to Colorless |
Density | 0.8010g/mL |
Flash Point | 3°C |
Assay Percent Range | 94% |
Packaging | AcroSeal™ Glass bottle |
Linear Formula | (CH3)2S·BH3 |
Quantity | 100 mL |
Fieser | 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50 |
Merck Index | 15, 1336 |
Show More |
Safety and Handling
Product Identifier
- Borane-methyl sulfide complex
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H301-Toxic if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'