Learn More
Borane-trimethylamine complex, 97%, Thermo Scientific Chemicals
Catalog Number p-71836
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
75-22-9
C3H12BN
72.95
MFCD00012420
WVMHLYQJPRXKLC-UHFFFAOYSA-N
Trimethylamine borane
trimethylamine borane
B.CN(C)C
75-22-9
C3H12BN
72.95
MFCD00012420
WVMHLYQJPRXKLC-UHFFFAOYSA-N
Trimethylamine borane
trimethylamine borane
B.CN(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 172.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)3N·BH3
Quantity 25 g
Fieser 01,1229; 05,708; 12,65
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ethanols,ethyl ether
Formula Weight 72.95
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Borane-trimethylamine complex, 98%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use to extinguish.