missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bordetella Pertussis Toxin Antibody (1280/204), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64514AF488
This item is not returnable.
View return policy
Description
Bordetella Pertussis Toxin Monoclonal specifically detects Bordetella Pertussis Toxin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Bordetella Pertussis Toxin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-64514 is reactive with Bordetella pertussis toxin in a simple ELISA. Reactivities with other organisms have not yet been determined. | |
| Bacteria | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 1280/204 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| pertussis toxin, pertussis toxin A protomer | |
| Native | |
| 0.1 mL | |
| Primary | |
| Protein A purified | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction