missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borealin Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22832AF350
This item is not returnable.
View return policy
Description
Borealin Polyclonal specifically detects Borealin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Borealin | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, DasraB, Dasra-B, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein | |
| Rabbit | |
| Affinity Purified | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger | |
| 55143.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CDCA8 | |
| Synthetic peptide made to an internal portion of human Borealin (within residues 200-250). [Swiss-ProtNo. Q53HL2] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction