missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borealin Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22832G
This item is not returnable.
View return policy
Description
Borealin Polyclonal specifically detects Borealin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Borealin | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Q53HL2 | |
| CDCA8 | |
| Synthetic peptide made to an internal portion of human Borealin (within residues 200-250). [Swiss-Prot# Q53HL2] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 92% identity with mouse and rat proteins. | |
| Store at 4C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, DasraB, Dasra-B, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein | |
| Rabbit | |
| 38 kDa | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger | |
| 55143 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction