missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borealin Antibody (OTI4G3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71988H
This item is not returnable.
View return policy
Description
Borealin Monoclonal specifically detects Borealin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Borealin | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CDCA8 | |
| Full length recombinant protein of human Borealin (NP_060571) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4G3 | |
| Western Blot, Immunohistochemistry | |
| BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, DasraB, Dasra-B, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein | |
| Mouse | |
| Affinity Purified | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger | |
| 55143.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction