Learn More
Boric Acid 1% Solution and Indicator For Kjeldahl N Det., Chem Lab
Brand: Chem Lab CL02.0204.5000
Additional Details : CAS : 10043-35-3 Weight : 5.00000kg
Specifications
Gray-Green | |
100°C | |
Polyethylene | |
Liquid |
0°C | |
1 g/mol | |
5 L | |
61.83 |
Safety and Handling
- Boric acid 1% solution + indicator
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Boric acid, Methanol