Learn More
WTW™ Boron B Reagent Test Kit
Click to view available options
No. of Tests:
60
Unit Size:
Each
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Specifications
| Analytes | Modified Winkler method |
| For Use With (Application) | For waste water, drinking water |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral photometers |
| Includes | Certificates |
| No. of Tests | 60 |
| Range | 0.050 - 0.800mg/I B |
| Test Method | Photometric |