Learn More
Jaytec Glass™ Borosilicate Glass Babcock Bottle
Product Code. 12358955
Change view
Click to view available options
Range:
0 to 50%
0 to 8%
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
12358955 0 to 50% Each
12348955 0 to 8% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12358955 Supplier Jaytec Glass™ Supplier No. MP.707
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Borosilicate Glass Babcock Bottle is used to determine the fat content of dairy samples.
  • Manufactured in accordance with BS.755, Part 1:1937
  • Borosilicate Glass
Specifications
Material Borosilicate Glass
Capacity (Metric) 9 g
Type Babcock Bottle
For Use With (Application) To measure fat content in cream
Range 0 to 50%