Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Funnels
Product Code. 10692472
Change view
Click to view available options
Porosity:
3
4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Porosity unitSize
10692472 3 Each
10548421 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10692472 Supplier Lenz Laborglasinstrumente™ Supplier No. 07200325
Add to basket
Add to basket
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Funnel is a laboratory funnel used for separating solids from liquids via the process of filtration. In order to achieve this, a cone-like shaped piece of filter paper is usually folded into a cone and placed within the funnel.
  • Conical shaped filter funnel
  • Made from borosilicate glass
Specifications
Capacity (Metric) 25 mL
Material Borosilicate Glass
Height (Metric) 100 mm
Diameter (Metric) Filter 25 mm
Porosity 3
Product Type Filter Funnel