Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
Product Code. 10475601
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 10475601
Brand: Lenz Laborglasinstrumente™ 04000073
Only null left
Add to basket
Add to basket
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
  • Without scale; with solid glass plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Stopper Material Polyethylene