Learn More
Karl Hecht™ Assistent™ Reduction and Expansion Adapters
Product Code. 10564042
Click to view available options
Standard Taper Joint No.:
Socket: 19/26
Socket: 29/32
Socket: 45/40
Cone Size:
14/23
19/26
24/29
29/32
45/50
Unit Size:
Each
Product Code. 10564042
Brand: Karl Hecht™ 42387010
Only null left
Add to basket
Add to basket
For the quick and secure connection of glass joints in the laboratory. Karl Hecht™ Assistent™ Reduction and Expansion Adapters are manufactured from durable DURAN™ borosilicate 3.3 glass.
  • Solid rim
  • Acc. to DIN 12257
  • Reinforced ground joint
Specifications
Cone Size 14/23
Material Borosilicate Glass
Product Type Duran sleeve
Quantity 1 Each
Standard Taper Joint No. Socket: 29/32