Learn More
IKA™ Borosilicate Glass Evaporating Flask For RV 10.86 Rotary Evaporators
€ 42.00 - € 201.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11855922
|
IKA™
0003740700 |
3000 mL |
€ 201.00
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11805932
|
IKA™
0003741200 |
50 mL |
€ 42.00
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Specifications
Borosilicate Glass | |
Evaporating Flask | |
1 Each |