Learn More
Robu™ Borosilicate Glass Filter Funnels
Brand:  Robu™ 22253
Additional Details : Weight : 0.33720kg
 View more versions of this product
Product Code. 16833046
  • € 255.00 / Pack of 10
Estimated Shipment: 04-06-2024
to see stock.
Specifications
55mm (OD); 25mm (ID)
8mm
Use in sample preparation processes and the filtration of small amounts
3
25 mL
Borosilicate Glass
55mm
Filter funnel