Description
- Resistant to aqueous solutions and solvents used in liquid chromatography
- Borosilicate glass tube
Specifications
Specifications
| Bed Volume | 120mL |
| Product Type | HiLoad Superose |
| Material | Borosilicate Glass tube |
| Support Type | Highly cross-linked agarose |
| Content And Storage | 4 to 30°C, 20% Ethanol |