missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Borosilicate Glass Laboratory Bottles, Blue Graduations

Product Code. 15314172
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15314172

Brand: Witeg™ 5 526 250 BL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains GL 45 screw thread. Witeg™ Borosilicate Glass Laboratory Bottles, Blue Graduations include a screw cap and polypropylene pouring ring.

  • Borosilicate Glass 3.3
  • GL 45 Screw Thread

Specifications

Capacity (Metric) 250 mL
Color Clear
For Use With (Application) Laboratory
Material Borosilicate Glass (3.3)
Closure Color Blue
Graduations Yes
Certifications/Compliance ISO 4796
Closure Included Yes
Mouth Wide
Product Type Wash Bottle
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.