Learn More
Kimble Chase™ Borosilicate Glass Plain Chromatography Column
Click to view available options
Column Capacity:
15 mL
25 mL
340 mL
440 mL
567 mL
589 mL
900 mL
965 mL
981 mL
1608 mL
1929 mL
2149 mL
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Ion exchange chromatography |
| Quantity | 1 Each |
| Diameter (Metric) Inner | 75 mm |
| Type | Plain Chromatography Column |
| USP Type | Type I |
| Column Capacity | 2149 mL |
| Length (Metric) | 500 mm |
| Material | Borosilicate Glass |
| Includes | With PTFE stopcock plug |