Learn More
Kimble Chase™ Borosilicate Glass Plain Chromatography Column
Product Code. 11867483
Click to view available options
Column Capacity:
15 mL
25 mL
340 mL
440 mL
567 mL
589 mL
900 mL
965 mL
981 mL
1608 mL
1929 mL
2149 mL
Unit Size:
Each
Product Code. 11867483
Brand: Kimble Chase™ 4205300630
Only null left
Specifications
For Use With (Application) Ion exchange chromatography
Quantity 1 Each
Diameter (Metric) Inner 64 mm
Type Plain Chromatography Column
USP Type Type I
Column Capacity 965 mL
Length (Metric) 300 mm
Material Borosilicate Glass
Includes With PTFE stopcock plug