Learn More
BRAND™ Borosilicate Glass Powder Funnel
Product Code. 10181531 Shop All Brand Products
Click to view available options
Diameter (Metric)Top:
60 mm
70 mm
80 mm
90 mm
100 mm
120 mm
Diameter (Metric) Stem:
18 mm
20 mm
22 mm
25 mm
30 mm
Unit Size:
Each
Product Code. 10181531
Brand: BRAND™ 146517
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
  • Manufactured from borosilicate glass.
  • Ground and fire-polished upper rim.
  • Short, wide stem optimal for funneling powder and other ground materials.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 90 mm
Diameter (Metric) Stem 25 mm
Material Borosilicate Glass