Learn More
Witeg™ Borosilicate Glass Powder Funnel with ST Cone
Witeg™ Borosilicate Glass Powder Funnels are available in a variety of cone tapers, angles and outer diameters.
Brand: Witeg™ 1 650 024
Additional Details : Weight : 0.04500kg
Description
- ST cone
- Borosilicate glass construction
- Available in a variety of tapers, angles, and diameters
Specifications
Funnel | |
Borosilicate Glass | |
24/29 |
80 mm | |
60° |