Learn More
Kimble Chase™ Borosilicate Glass Serum Vials without Closures
Kimble Chase™ Borosilicate Glass Serum Vials without Closures are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples, and more.
€ 540.00 - € 1880.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10023444
|
Kimble Chase™
62121D-5 |
5 mL |
€ 540.00
Pack of 864 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
12827690
|
Kimble Chase™
62113U-2 |
2 mL |
€ 1880.00
Pack of 4640 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
Description
- Special design provides extra strength for freeze-drying applications
- Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
- Special blowback design provides ease of use for lyophillization stoppers
- Autoclavable
- Aluminum seals, stoppers and septa are available separately
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Borosilicate Glass (Type 1 Class A) |