Learn More
Karl Hecht™ Bottle, Narrow Neck with Glass Stopper
Product Code. p-7115713
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
Color:
Amber
Clear
Unit Size:
Each
Product Code. 10409981
Brand: Karl Hecht™ 41303018
Only null left
Add to basket
Add to basket
Round-shouldered chemical/reagent bottle. Karl Hecht™ Bottle, Narrow Neck with Glass Stopper includes a ground-in flat glass stopper.
  • Narrow neck.
  • Round shoulder.
  • Ground in flat glass stopper.
  • Available in clear glass or amber glass.
Specifications
Closure Material Glass
Color Amber
Capacity (Metric) 250 mL
Material Glass
Neck Style Narrow
Product Type Bottle
Shape Round Shoulder
Closure Type Stopper