Learn More
Karl Hecht™ Bottles, Wide Neck with Glass Stopper
Product Code. 10573842
Change view
Click to view available options
:
CapacityMetric: 500mL, Material: Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10573842 CapacityMetric: 500mL, Material: Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10573842 Supplier Karl Hecht™ Supplier No. 41311024
Round-shouldered chemical/reagent bottle. Karl Hecht™ Bottle, Wide Neck with Glass Stopper includes a ground-in flat glass stopper.
  • Wide neck.
  • Round shoulder.
  • Ground in flat glass stopper.
Specifications
Closure Material Glass
Color Clear
Capacity (Metric) 500 mL
Material Glass
Neck Style Wide
Product Type Bottle
Shape Round Shoulder
Closure Type Stopper