missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Fetuin (custom product)

Bovine Fetuin (custom product)

Brand:  Gibco™ 10344034

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11558836

  • € 2335.00 / 10g
Estimated Shipment: 01-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • GIBCO Custom Product
  • Custom Media Inquiry Form
Specifications

Specifications

10 g
Store in refrigerator (2–8°C).
Gibco™
Bovine
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers