missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine FGF basic/FGF2/bFGF Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AB-33-NA
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Bovine samples. It is validated for Western Blot, Neutralization.
Specifications
| FGF basic/FGF2 | |
| Polyclonal | |
| Unconjugated | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2247 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1.5-4.5 ug/mL | |
| FGF2 | |
| Bovine brain-derived FGF basic/FGF2 | |
| 1 mg | |
| Primary | |
| Detects bovine FGF basic/FGF2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with FGF acidic and recombinant human (rh) beta-ECGF is observed and less than 1% cross-reactivity with rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-7, recombinant mouse (rm) FGF-8b, rmFGF-8c, rhFGF-9, rhFGF-10, rmFGF-15, rhFGF-17, and rhFGF-18 is observed. Neutralizes the biological activity of bovine FGF basic/FGF2 and will also neutralize the biological activity of recombinant human FGF basic/FGF2. | |
| Bovine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction