missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine IFN-gamma Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2300
This item is not returnable.
View return policy
Description
IFN-gamma Polyclonal specifically detects IFN-gamma in Bovine samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IFN-gamma | |
| Polyclonal | |
| Unconjugated | |
| NP_776511 | |
| IFNG | |
| E. coli-derived recombinant bovine IFN-gamma Gln24-Thr166 Accession # NP_776511 | |
| 100 μg | |
| Primary | |
| Detects bovine IFN-gamma in direct ELISAs and Western blots. | |
| Bovine | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 10-20 ug/mL | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3458 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction