missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine Papilloma Virus Monoclonal Antibody (I24F)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17407
This item is not returnable.
View return policy
Description
The MA1-7407 antibody reacts with Bovine Papillomavirus in viral and infected tissue samples. MA1-7407 has been successfully used in ELISA applications. The MA1-7407 antibody was raised against a BPV preparation.
Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
Specifications
| Bovine Papilloma Virus | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| I24F | |
| Unconjugated | |
| BPV | |
| BPV preparation. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction