missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (3G3A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2195
This item is not returnable.
View return policy
Description
Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Bovine samples. It is validated for Western Blot, Simple Western, ELISA.
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Simple Western, ELISA 1:10000 | |
| ALB | |
| Bovine serum albumin. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 280717 | |
| Bovine | |
| Purified |
| Western Blot, ELISA | |
| 3H6 | |
| Unconjugated | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| Bovine serum albumin (BSA). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction